Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAR7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343726.1 | NP_001331820.1 | 6 - 7 | 1761 | 126 | |
NM_001343727.1 | NP_001331821.1 | 7 - 8 | 1432 | 126 | ||
NM_001343728.1 | NP_001331818.1 | 8 - 9 | 1325 | 126 | ||
NM_001343729.1 | NP_001331819.1 | 7 - 8 | 1224 | 126 | ||
NM_001343730.1 | NP_001331816.1 | 6 - 7 | 1279 | 126 | ||
NM_001343731.1 | NP_001331817.1 | 4 - 5 | 806 | 126 | ||
NM_122147.2 | NP_197634.1 | 7 - 8 | 973 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |