Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHX9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122197.1 | NP_197682.1 | 1 - 2 | 250 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |