Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122208.4 | NP_197693.1 | 3 - 4 | 1001 | 77 | |
GenBank mRNA | AJ486937.1 | - | 3 - 4 | 916 | 77 | |
AJ486942.1 | - | 3 - 4 | 916 | 77 | ||
AJ486945.1 | - | 3 - 4 | 916 | 77 | ||
AJ486946.1 | - | 3 - 4 | 916 | 77 | ||
AY099549.1 | - | 3 - 4 | 997 | 77 | ||
BT008874.1 | - | 3 - 4 | 916 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |