Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343816.1 | NP_001331492.1 | 2 - 3 | 1969 | 103 | |
NM_001343817.1 | NP_001331491.1 | 2 - 3 | 1969 | 103 | ||
NM_001343818.1 | NP_001331490.1 | 3 - 4 | 1425 | 103 | ||
NM_122263.3 | NP_197747.1 | 3 - 4 | 1563 | 103 | ||
GenBank mRNA | BT002944.1 | - | 3 - 4 | 1347 | 103 | |
BT004380.1 | - | 2 - 3 | 1256 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |