Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLAH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122308.5 | NP_197791.1 | 4 - 5 | 1798 | 98 | |
GenBank mRNA | BT015364.1 | - | 4 - 5 | 1630 | 98 | |
BT015721.1 | - | 3 - 4 | 1488 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |