Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122314.3 | NP_197797.1 | 2 - 3 | 606 | 92 | |
GenBank mRNA | BT020526.1 | - | 2 - 3 | 606 | 92 | |
BT021931.1 | - | 2 - 3 | 507 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |