Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036864.1 | NP_001031941.1 | 4 - 5 | 917 | 58 | |
NM_122504.5 | NP_197975.3 | 4 - 5 | 926 | 58 | ||
GenBank mRNA | AK226466.1 | - | 4 - 5 | 759 | 58 | |
X73417.1 | - | 768 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |