Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYP41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036873.2 | NP_001031950.1 | 1 - 2 | 685 | 75 | |
NM_001343989.1 | NP_001331924.1 | 1 - 2 | 424 | 75 | ||
NM_122580.4 | NP_198050.1 | 1 - 2 | 685 | 75 | ||
GenBank mRNA | AF067789.2 | - | 1 - 2 | 278 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |