Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZYP1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332545.1 | NP_001323354.1 | 16 - 17 | 2681 | 78 | |
NM_102078.3 | NP_564164.1 | 17 - 18 | 2649 | 78 | ||
GenBank mRNA | DQ028765.1 | - | 17 - 18 | 2583 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |