Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LPR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102149.4 | NP_173714.2 | 2 - 3 | 1585 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |