Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102181.1 | NP_173746.1 | 2 - 3 | 393 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |