Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSLA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332636.1 | NP_001320353.1 | 6 - 7 | 1204 | 73 | |
NM_102254.3 | NP_173818.1 | 6 - 7 | 1259 | 73 | ||
GenBank mRNA | KJ676988.1 | - | 6 - 7 | 1009 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |