Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEE8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102341.1 | NP_173902.1 | 3 - 4 | 352 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |