Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PFT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084127.1 | NP_001077596.1 | 7 - 8 | 1075 | 107 | |
NM_102365.5 | NP_173925.3 | 8 - 9 | 1261 | 107 | ||
GenBank mRNA | AY170377.1 | - | 8 - 9 | 892 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |