Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYCT1;3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332773.1 | NP_001321314.1 | 5 - 6 | 1426 | 73 | |
NM_001332774.1 | NP_001321315.1 | 4 - 5 | 1536 | 73 | ||
NM_001332775.1 | NP_001321316.1 | 5 - 6 | 823 | 73 | ||
NM_001332776.1 | NP_001321317.1 | 5 - 6 | 1205 | 73 | ||
NM_001332777.1 | NP_001321318.1 | 3 - 4 | 1100 | 73 | ||
NM_102528.4 | NP_174084.1 | 5 - 6 | 873 | 73 | ||
GenBank mRNA | AF344323.1 | - | 4 - 5 | 617 | 73 | |
AK117692.1 | - | 5 - 6 | 823 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |