Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP65-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332791.1 | NP_001322317.1 | 3 - 4 | 700 | 172 | |
NM_102557.3 | NP_174113.2 | 3 - 4 | 993 | 172 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |