Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334601.1 | NP_001322105.1 | 1 - 2 | 1872 | 75 | |
NM_106032.5 | NP_177514.1 | 2 - 3 | 2056 | 75 | ||
GenBank mRNA | AF004215.1 | - | 2 - 3 | 1837 | 75 | |
AY070044.1 | - | 2 - 3 | 1887 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |