Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RLP16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334636.1 | NP_001321481.1 | 2 | 1853 | 119 | |
NM_106080.2 | NP_177560.2 | 1 | 1978 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |