Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106086.5 | NP_177566.3 | 2 - 3 | 662 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |