Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACBP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344021.1 | NP_001331944.1 | 9 - 10 | 1125 | 101 | |
NM_001344022.1 | NP_001331946.1 | 9 - 10 | 1125 | 101 | ||
NM_001344023.1 | NP_001331945.1 | 9 - 10 | 956 | 101 | ||
NM_122645.5 | NP_198115.2 | 9 - 10 | 982 | 101 | ||
GenBank mRNA | AY093156.1 | - | 9 - 10 | 956 | 101 | |
BT008399.1 | - | 9 - 10 | 878 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |