Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344066.1 | NP_001330383.1 | 11 - 12 | 1426 | 64 | |
NM_001344067.1 | NP_001330382.1 | 12 - 13 | 943 | 64 | ||
NM_122712.3 | NP_198181.1 | 12 - 13 | 1153 | 64 | ||
GenBank mRNA | AY090360.1 | - | 12 - 13 | 942 | 64 | |
BT015415.1 | - | 11 - 12 | 690 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |