Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: iqd33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122958.4 | NP_568529.1 | 1 - 2 | 514 | 110 | |
GenBank mRNA | AK175704.1 | - | 1 - 2 | 145 | 110 | |
AY088904.1 | - | 1 - 2 | 135 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |