Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122966.3 | NP_568532.1 | 11 - 12 | 3260 | 84 | |
GenBank mRNA | AB046869.1 | - | 11 - 12 | 2792 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |