Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YAK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036893.2 | NP_001031970.1 | 9 - 10 | 1593 | 134 | |
NM_001344125.1 | NP_001332507.1 | 9 - 10 | 1071 | 134 | ||
NM_122989.4 | NP_198447.2 | 9 - 10 | 1617 | 134 | ||
GenBank mRNA | AK230350.1 | - | 9 - 10 | 1535 | 134 | |
AY093090.1 | - | 9 - 10 | 1593 | 134 | ||
BT010549.1 | - | 9 - 10 | 1026 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |