Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203506.1 | NP_001190435.1 | 2 - 3 | 406 | 91 | |
NM_123227.6 | NP_198682.3 | 2 - 3 | 1374 | 91 | ||
GenBank mRNA | BT024514.1 | - | 2 - 3 | 318 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |