Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203506.1 | NP_001190435.1 | 6 - 7 | 742 | 78 | |
NM_123227.6 | NP_198682.3 | 6 - 7 | 1710 | 78 | ||
GenBank mRNA | BT024514.1 | - | 6 - 7 | 654 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |