Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIG2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344301.1 | NP_001330763.1 | 1 - 2 | 179 | 115 | |
NM_123334.3 | NP_198788.1 | 1 - 2 | 179 | 115 | ||
GenBank mRNA | BT030037.1 | - | 1 - 2 | 124 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |