Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344355.1 | NP_001332644.1 | 14 - 15 | 8015 | 63 | |
NM_001344356.1 | NP_001332643.1 | 14 - 15 | 8012 | 63 | ||
NM_001344357.1 | NP_001332642.1 | 14 - 15 | 8012 | 63 | ||
NM_123447.3 | NP_198898.2 | 14 - 15 | 8012 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |