Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UVH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036914.1 | NP_001031991.1 | 3 - 4 | 1207 | 73 | |
NM_001344387.1 | NP_001330263.1 | 3 - 4 | 1673 | 73 | ||
NM_001344388.1 | NP_001330264.1 | 3 - 4 | 1207 | 73 | ||
NM_123480.4 | NP_198931.1 | 3 - 4 | 1673 | 73 | ||
GenBank mRNA | AF089003.1 | - | 3 - 4 | 1175 | 73 | |
AF160500.1 | - | 3 - 4 | 1269 | 73 | ||
AY140076.1 | - | 3 - 4 | 1207 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |