Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123524.4 | NP_198975.1 | 4 - 5 | 800 | 73 | |
GenBank mRNA | AY035169.1 | - | 4 - 5 | 653 | 73 | |
AY057638.1 | - | 4 - 5 | 699 | 73 | ||
AY150449.1 | - | 4 - 5 | 599 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |