Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scpl41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344440.1 | NP_001318730.1 | 4 - 5 | 684 | 67 | |
NM_123588.4 | NP_199038.1 | 3 - 4 | 844 | 67 | ||
GenBank mRNA | BT002948.1 | - | 3 - 4 | 477 | 67 | |
BT004386.1 | - | 2 - 3 | 346 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |