Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123695.3 | NP_199143.1 | 1 - 2 | 396 | 73 | |
GenBank mRNA | AB493775.1 | - | 1 - 2 | 258 | 73 | |
AK176443.1 | - | 1 - 2 | 376 | 73 | ||
BT010424.1 | - | 1 - 2 | 258 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |