Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETFBETA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036925.2 | NP_001032002.1 | 3 - 4 | 487 | 77 | |
NM_001085230.2 | NP_001078699.1 | 3 - 4 | 477 | 77 | ||
NM_001344482.1 | NP_001332757.1 | 3 - 4 | 568 | 77 | ||
NM_123709.4 | NP_199156.1 | 3 - 4 | 504 | 77 | ||
GenBank mRNA | AF446867.1 | - | 3 - 4 | 429 | 77 | |
AK317065.1 | - | 3 - 4 | 470 | 77 | ||
AK317388.1 | - | 3 - 4 | 466 | 77 | ||
AY045657.1 | - | 3 - 4 | 478 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |