Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TZP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344491.1 | NP_001330182.1 | 2 - 3 | 1042 | 70 | |
NM_001344492.1 | NP_001330183.1 | 3 - 4 | 871 | 70 | ||
NM_001344493.1 | NP_001330181.1 | 3 - 4 | 577 | 70 | ||
NM_123729.3 | NP_199176.1 | 3 - 4 | 592 | 70 | ||
GenBank mRNA | BT003987.1 | - | 3 - 4 | 570 | 70 | |
BT005699.1 | - | 2 - 3 | 441 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |