Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123752.4 | NP_568630.1 | 2 - 3 | 348 | 68 | |
GenBank mRNA | BT000432.1 | - | 2 - 3 | 143 | 68 | |
BT006566.1 | - | 1 - 2 | 107 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |