Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344531.1 | NP_001332549.1 | 8 - 9 | 1816 | 69 | |
NM_123774.4 | NP_199220.1 | 8 - 9 | 1546 | 69 | ||
GenBank mRNA | AF093753.1 | - | 8 - 9 | 1376 | 69 | |
AF135155.1 | - | 8 - 9 | 1424 | 69 | ||
AY039951.1 | - | 8 - 9 | 1494 | 69 | ||
AY048257.1 | - | 8 - 9 | 1485 | 69 | ||
AY079384.1 | - | 8 - 9 | 1376 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |