Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344590.1 | NP_001332060.1 | 20 - 21 | 1839 | 72 | |
NM_001344591.1 | NP_001332059.1 | 20 - 21 | 1757 | 72 | ||
NM_123882.5 | NP_199327.4 | 20 - 21 | 1757 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |