Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344604.1 | NP_001318742.1 | 5 - 6 | 4842 | 89 | |
NM_123894.4 | NP_199339.2 | 6 - 7 | 3754 | 89 | ||
GenBank mRNA | AK226456.1 | - | 6 - 7 | 3755 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |