Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KASI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036941.1 | NP_001032018.1 | 7 - 8 | 1292 | 60 | |
NM_001203550.1 | NP_001190479.1 | 6 - 7 | 1512 | 60 | ||
NM_123998.3 | NP_199441.1 | 6 - 7 | 1561 | 60 | ||
GenBank mRNA | AY037261.1 | - | 6 - 7 | 1451 | 60 | |
AY087843.1 | - | 6 - 7 | 1426 | 60 | ||
AY094005.1 | - | 6 - 7 | 1351 | 60 | ||
AY123979.1 | - | 6 - 7 | 1455 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |