Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124005.5 | NP_199447.1 | 1 - 2 | 613 | 118 | |
GenBank mRNA | AF404855.1 | - | 1 - 2 | 458 | 118 | |
AY063916.1 | - | 1 - 2 | 599 | 118 | ||
AY096426.1 | - | 1 - 2 | 458 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Arabidopsis thaliana Probe spans exons |