Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBP21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344697.1 | NP_001330462.1 | 4 - 5 | 1127 | 117 | |
NM_124044.2 | NP_568667.1 | 4 - 5 | 1193 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |