Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203563.2 | NP_001190492.1 | 14 - 15 | 2017 | 85 | |
NM_124236.3 | NP_199671.1 | 14 - 15 | 1868 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |