Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GEX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344816.1 | NP_001330856.1 | 12 - 13 | 2605 | 106 | |
NM_001344817.1 | NP_001330859.1 | 12 - 13 | 2605 | 106 | ||
NM_001344819.1 | NP_001330858.1 | 11 - 12 | 2269 | 106 | ||
NM_001344820.1 | NP_001330862.1 | 11 - 12 | 2269 | 106 | ||
NM_001344821.1 | NP_001330861.1 | 9 - 10 | 1777 | 106 | ||
NM_001344822.1 | NP_001330857.1 | 9 - 10 | 1777 | 106 | ||
NM_124292.3 | NP_199726.3 | 12 - 13 | 2576 | 106 | ||
GenBank mRNA | AB743888.1 | - | 12 - 13 | 2694 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |