Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085271.1 | NP_001078740.1 | 3 - 4 | 685 | 93 | |
NM_001125934.1 | NP_001119406.1 | 2 - 3 | 431 | 93 | ||
NM_001344883.1 | NP_001331546.1 | 3 - 4 | 321 | 93 | ||
NM_124403.2 | NP_199835.2 | 3 - 4 | 694 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |