Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PBS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344954.1 | NP_001331373.1 | 2 - 3 | 1044 | 83 | |
NM_124549.8 | NP_568762.6 | 3 - 4 | 333 | 83 | ||
GenBank mRNA | AF192262.1 | - | 3 - 4 | 242 | 83 | |
AY088919.1 | - | 3 - 4 | 252 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |