Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124589.3 | NP_200023.1 | 6 - 7 | 737 | 73 | |
GenBank mRNA | BT030092.1 | - | 6 - 7 | 582 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |