Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LTI78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124610.3 | NP_200044.1 | 2 - 3 | 444 | 76 | |
GenBank mRNA | L22567.1 | - | 2 - 3 | 310 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |