Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNGC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124692.3 | NP_200125.1 | 3 - 4 | 999 | 108 | |
GenBank mRNA | AF370139.1 | - | 3 - 4 | 999 | 108 | |
BT000991.1 | - | 2 - 3 | 606 | 108 | ||
Y16327.1 | - | 3 - 4 | 910 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |