Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203600.1 | NP_001190529.1 | 10 - 11 | 2107 | 102 | |
NM_001203601.1 | NP_001190530.1 | 10 - 11 | 2107 | 102 | ||
NM_124725.5 | NP_200158.2 | 10 - 11 | 2122 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |