Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAMI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345079.1 | NP_001331145.1 | 5 - 6 | 1589 | 77 | |
NM_124791.3 | NP_568804.1 | 6 - 7 | 1036 | 77 | ||
GenBank mRNA | BT025794.1 | - | 6 - 7 | 739 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |